Skip to Content
Merck
All Photos(3)

Documents

678937

Sigma-Aldrich

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine

97%

Synonym(s):

TBTA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H30N10
CAS Number:
Molecular Weight:
530.63
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: ligand
reaction type: click chemistry

mp

132-143 °C

storage temp.

−20°C

SMILES string

C(N(Cc1cn(Cc2ccccc2)nn1)Cc3cn(Cc4ccccc4)nn3)c5cn(Cc6ccccc6)nn5

InChI

1S/C30H30N10/c1-4-10-25(11-5-1)16-38-22-28(31-34-38)19-37(20-29-23-39(35-32-29)17-26-12-6-2-7-13-26)21-30-24-40(36-33-30)18-27-14-8-3-9-15-27/h1-15,22-24H,16-21H2

InChI key

WKGZJBVXZWCZQC-UHFFFAOYSA-N

Application

A polytriazolylamine ligand which stabilizes Cu(I) towards disproportionation and oxidation thus enhancing its catalytic effect in the azide-acetylene cycloaddition.

related product

Product No.
Description
Pricing

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Frank Iversen et al.
Theranostics, 3(3), 201-209 (2013-03-09)
Some of the main concerns with in vivo application of naked small interfering RNA are rapid degradation and urinary excretion resulting in a short plasma half-life. In this study we investigated how conjugation of polyethylene glycol (PEG) with variable chain
Brent R Martin et al.
Nature methods, 9(1), 84-89 (2011-11-08)
The reversible thioester linkage of palmitic acid on cysteines, known as protein S-palmitoylation, facilitates the membrane association and proper subcellular localization of proteins. Here we report the metabolic incorporation of the palmitic acid analog 17-octadecynoic acid (17-ODYA) in combination with
Timothy R Chan et al.
Organic letters, 6(17), 2853-2855 (2004-08-28)
Polytriazolylamines were synthesized by the copper(I)-catalyzed ligation of azides and alkynes. The C3-symmetric derivative, TBTA, was shown to be a powerful stabilizing ligand for copper(I), protecting it from oxidation and disproportionation, while enhancing its catalytic activity.
Xiangyu Zhang et al.
Therapeutic advances in medical oncology, 12, 1758835920947976-1758835920947976 (2020-10-01)
Aspirin has recently emerged as an anticancer drug, but its therapeutic effect on lung cancer has been rarely reported, and the mechanism of action is still unclear. Long-term use of celecoxib in large doses causes serious side effects, and it
Laurie D Cohen et al.
PloS one, 8(5), e63191-e63191 (2013-05-10)
Chemical synapses contain multitudes of proteins, which in common with all proteins, have finite lifetimes and therefore need to be continuously replaced. Given the huge numbers of synaptic connections typical neurons form, the demand to maintain the protein contents of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service