900694
10-(3,5-Dimethoxyphenyl)-9-mesityl-1,3,6,8-tetramethoxyacridin-10-ium tetrafluoroborate
≥93%
Synonym(s):
10-(3,5-dimethoxyphenyl)-1,3,6,8-tetramethoxy-9-(2,4,6-trimethylphenyl)-acridinium tetrafluoroborate
About This Item
Recommended Products
Quality Level
Assay
≥93%
form
powder or crystals
reaction suitability
core: acridinium
reagent type: catalyst
reaction type: Photocatalysis
mp
242-247 °C
photocatalyst activation
460 nm
SMILES string
O(P(OC[C@@H](COC(C(C(C(C(C(C(C(C(C(C(C(C(C(C(C([2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])([2H])[2H])=O)OC(CCCCCCC/C=C\CCCCCCCC)=O)(=O)O)[C@H]1[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@@H]1O.N
InChI
1S/C43H81O13P.H3N/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(45)55-35(34-54-57(51,52)56-43-41(49)39(47)38(46)40(48)42(43)50)33-53-36(44)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,35,38-43,46-50H,3-16,19-34H2,1-2H3,(H,51,52);1H3/b18-17-;/t35-,
InChI key
NPOZNLWSCICHRX-ZXXUQEDFSA-N
Application
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service