900421
9-Mesityl-3,6-di-tert-butyl-10-phenylacridinium tetrafluoroborate
≥95%
Synonym(s):
3,6-Bis(1,1-dimethylethyl)-10-phenyl-9-(2,4,6-trimethylphenyl)-acridinium tetrafluoroborate, 3,6-Di-tert-butyl-9-(2,4,6-trimethylphenyl)-10-phenylacridinium tetrafluoroborate
About This Item
Quality Level
Assay
≥95%
form
powder or crystals
reaction suitability
core: acridinium
reagent type: catalyst
reaction type: Photocatalysis
mp
277.2 °C
photocatalyst activation
455 nm
SMILES string
[F-][B+3]([F-])([F-])[F-].C=1C=CC(=CC1)[N+]=2C=3C=C(C=CC3C(=C4C=CC(=CC42)C(C)(C)C)C=5C(=CC(=CC5C)C)C)C(C)(C)C
InChI
InChI=1S/C36H40N.BF4/c1-23-19-24(2)33(25(3)20-23)34-29-17-15-26(35(4,5)6)21-31(29)37(28-13-11-10-12-14-28)32-22-27(36(7,8)9)16-18-30(32)34;2-1(3,4)5/h10-22H,1-9H3;/q+1;-1
InChI key
YUNSHKYOOCWQRB-UHFFFAOYSA-N
Related Categories
Application
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service