Skip to Content
Merck
All Photos(1)

Key Documents

86884

Sigma-Aldrich

Tetrabutylammonium nitrite

≥97.0% (NT)

Synonym(s):

N,N,N-tributyl-1-butanaminium nitrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(NO2)
CAS Number:
Molecular Weight:
288.47
Beilstein:
6285720
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (NT)

form

crystals

SMILES string

[O-]N=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.HNO2/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2-1-3/h5-16H2,1-4H3;(H,2,3)/q+1;/p-1

InChI key

SHRKDVQQQPFSIY-UHFFFAOYSA-M

Related Categories

Other Notes

Reagent for the synthesis of aliphatic nitrocompounds from bromides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Munz et al.
Liebigs Ann. Chem., 628-628 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service