Skip to Content
Merck
All Photos(1)

Key Documents

87741

Sigma-Aldrich

Tetramethylammonium hydroxide pentahydrate

≥95.0% (T)

Synonym(s):

TMAH Pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(OH) · 5H2O
CAS Number:
Molecular Weight:
181.23
Beilstein:
3714235
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.1 (vs air)

Assay

≥95.0% (T)

form

crystals

autoignition temp.

878 °F

expl. lim.

36 %

impurities

≤0.1% halides (as chloride)

pH

(14 at ca.1,000 g/l at 20 °C (68 °F))

mp

67-70 °C (lit.)

solubility

soluble (2,200 g/l at 15 °C (59 °F))

anion traces

sulfate (SO42-): ≤1000 mg/kg

functional group

amine

SMILES string

O.O.O.O.O.[OH-].C[N+](C)(C)C

InChI

1S/C4H12N.6H2O/c1-5(2,3)4;;;;;;/h1-4H3;6*1H2/q+1;;;;;;/p-1

InChI key

MYXKPFMQWULLOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetramethylammonium hydroxide pentahydrate is a clathrate hydrate, which shows high proton conductivity. It can readily uptake carbon dioxide. It is a promising candidate for energy devices, gas separation, and gas storage applications.

Application

Tetramethylammonium hydroxide pentahydrate is a clathrate hydrate, which shows high proton conductivity. It can readily uptake carbon dioxide. It is a promising candidate for energy devices, gas separation, and gas storage applications.
Tetramethylammonium hydroxide pentahydrate is a clathrate hydrate, which shows high proton conductivity. It can readily uptake carbon dioxide. It is a promising candidate for energy devices, gas separation, and gas storage applications.

Tetramethylammonium hydroxide pentahydrate may be used in the preparation of a mesoporous molecular sieve belonging to the MCM-41 (mobil composition of matter No. 41) category with silica to (tetrahedral) aluminum ratio as low as 8.5:1.

Caution

material is sensitive vs air/moisture. filling, handling and storage under Argon!

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1 Dermal - STOT SE 1

Target Organs

Central nervous system, Liver,thymus

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a mesoporous MCM-41 material with high levels of tetrahedral aluminium.
Schmidt R, et al.
Journal of the Chemical Society. Chemical Communications, (12), 1493-1494 (1994)
High conductivity and phase transitions of tetramethylammonium hydroxide pentahydrate,(CH3) 4NOH? 5H2O.
Kuriyama N, et al.
Solid State Ionics, 40, 906-909 (1990)
NMR studies of molecular motion in tetramethylammonium hydroxide pentahydrate.
Ratcliffe CI, et al.
J. Incl. Phenom. Mol. Recog. Chem., 8(1-2), 159-175 (1990)
Proton Conduction of Tetramethylammonium Hydroxide Pentahydrate,(CH 3) 4 NOH? 5 H 2 O, and Its Application to Nickel?Metal Hydride Battery.
Kuriyama N, et al.
Journal of the Electrochemical Society, 137(1), 355-356 (1990)
Wei Shen et al.
Chemical communications (Cambridge, England), 49(73), 8114-8116 (2013-08-08)
A simple and low-cost colorimetric assay utilizing ferrofluidic nanoparticulate probes (FNPs) and a ligase for single-nucleotide polymorphism genotyping is described. Excellent sensitivity and selectivity were accomplished through the engagement of the FNPs and a ligase chain reaction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service