Skip to Content
Merck
All Photos(1)

Key Documents

797537

Sigma-Aldrich

PhenoFluor Mix

Synonym(s):

N,N?′-1,3-bis(2,6-diisopropylphenyl)chloroimidazolium chloride and CsF mixture (1:2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

mp

228.65 °C

SMILES string

[Cs]F.Cl[C+]1=[N](C2=C(C(C)C)C=CC=C2C(C)C)C=CN1C3=C(C(C)C)C=CC=C3C(C)C.[Cl-]

InChI

1S/C27H36ClN2.ClH.Cs.FH/c1-17(2)21-11-9-12-22(18(3)4)25(21)29-15-16-30(27(29)28)26-23(19(5)6)13-10-14-24(26)20(7)8;;;/h9-20H,1-8H3;1H;;1H/q+1;;+1;/p-2

InChI key

AHNDOFVAHQKSBA-UHFFFAOYSA-L

General description

PhenoFluor Mix is a mixture of N,N′-1,3-bis(2,6-diisopropylphenyl)chloroimidazolium chloride and CsF in 1:2 weight ratio.

Application

PhenoFluor Mix is a benchtop stable alternative to PhenoFluor, which is a reagent for the deoxyfluorination of phenols, including electron-deficient, electron-rich, and heterocyclic phenols. Deoxyfluorination with PhenoFluor Mix is operationally simple and scalable.

Other Notes

Mix well, before use. Material can separate when not in use.

Legal Information

PhenoFluor is a trademark of SciFluor Life Sciences, LLC
PhenoFluor is a trademark of SciFluor Life Sciences, LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Kidney,Adrenal gland, Respiratory system

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

PhenoFluorMix: practical chemoselective deoxyfluorination of phenols.
Fujimoto T and Ritter T
Organic Letters, 17(3), 544-547 (2015)
Facile C-F Bond Formation through a Concerted Nucleophilic Aromatic Substitution Mediated by the PhenoFluor Reagent.
Neumann CN and Ritter T
Accounts of Chemical Research, 50(11), 2822-2833 (2017)
AlkylFluor: Deoxyfluorination of Alcohols.
Goldberg NW, et al.
Organic Letters, 18(23), 6102-6104 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service