Skip to Content
Merck
All Photos(1)

Documents

793329

Sigma-Aldrich

Biotin-benzyl-tetrazine

95%

Synonym(s):

Biotin-benzyl-tetrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N7O2S
Molecular Weight:
413.50
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

reaction suitability

reagent type: cross-linking reagent

storage temp.

−20°C

SMILES string

O=C(CCCC[C@@H](SC1)[C@@]2([H])[C@]1([H])NC(N2)=O)NCC(C=C3)=CC=C3C4=NN=CN=N4

InChI

1S/C19H23N7O2S/c27-16(4-2-1-3-15-17-14(10-29-15)23-19(28)24-17)20-9-12-5-7-13(8-6-12)18-25-21-11-22-26-18/h5-8,11,14-15,17H,1-4,9-10H2,(H,20,27)(H2,23,24,28)/t14-,15?,17-/m1/s1

InChI key

GBJXXSUDJUYIIH-XZDBBAETSA-N

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service