Skip to Content
Merck
All Photos(1)

Documents

779334

Sigma-Aldrich

β-Cyclodextrin

Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, ≥98.0% (HPLC)

Synonym(s):

Cavamax® W7 Pharma, beta-Cyclodextrin, Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H70O35
CAS Number:
Molecular Weight:
1134.98
Beilstein:
78623
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

Produced by Wacker Chemie AG, Burghausen, Germany, Life Science

Assay

≥98.0% (HPLC)

form

powder

optical activity

[α]/D 160 to 164°
[α]20/D +162±3°, c = 1.5% in H2O

impurities

≤0.2% reducing substances
≤0.25% α- and γ-cyclodextrin (each)
≤0.5% related substances
≤5 ppm heavy metals
≤5 ppm residual solvents

ign. residue

≤0.10%

loss

≤14.0% loss on drying

pH

5.0-8.0 (1% in solution)

mp

290-300 °C (dec.) (lit.)

solubility

solution: 1%, clear

absorption

≤0.05 at 350-750 nm in solution at 1%
≤0.1 at 230-350 nm in solution at 1%

suitability

corresponds for identity

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C42H70O35/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51/h8-63H,1-7H2/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-/m1/s1

InChI key

WHGYBXFWUBPSRW-FOUAGVGXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For R&D use

Legal Information

Cavasol is a registered trademark of Wacker Chemie AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanane Messiad et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 926, 21-27 (2013-03-30)
Piceatannol is one of resveratrol derivatives having health promoting potential. However, its low water-solubility and bioavailability could limit its use in both food and pharmaceutical fields. The aim of this work is the study of piceatannol complexation by β-cyclodextrin (β-CD)
Shuye Wang et al.
Journal of chromatography. A, 1277, 84-92 (2013-01-12)
Methoxyethylamine monosubstituted β-cyclodextrin, mono-6(A)-(2-methoxyethyl-1-ammonium)-6(A)-β-cyclodextrin chloride (MEtAMCD), is synthesized and analytically characterized. Bearing a methoxy group in cyclodextrin rim, MEtAMCD exhibits outstanding enantioselectivities toward ampholytic and acidic racemates in capillary electrophoresis. Driven by inclusion complexation, electrostatic interactions and/or hydrogen bonding, the
Xu Zhang et al.
Chemical communications (Cambridge, England), 49(8), 825-827 (2012-12-14)
This work revealed that β-cyclodextrin was attached onto the surface of TiO(2) predominately by its secondary ring side, which caused paradoxical functions of β-cyclodextrin in the photodegradation of the four bisphenols. The equilibrium between the guest adsorbed through β-cyclodextrin onto
L Siman et al.
Physical review letters, 109(24), 248103-248103 (2013-02-02)
Binding of ligands to DNA can be studied by measuring the change of the persistence length of the complex formed, in single-molecule assays. We propose a methodology for persistence length data analysis based on a quenched disorder statistical model and
Erik T Jansson et al.
Molecular pain, 9, 1-1 (2013-01-03)
The TRPV1 ion channel is expressed in nociceptors, where pharmacological modulation of its function may offer a means of alleviating pain and neurogenic inflammation processes in the human body. The aim of this study was to investigate the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service