Skip to Content
Merck
All Photos(1)

Documents

28705

Sigma-Aldrich

α-Cyclodextrin

purum, ≥98.0% (HPLC)

Synonym(s):

alpha-Cyclodextrin, α-Schardinger dextrin, Cyclohexaamylose, Cyclomaltohexaose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60O30
CAS Number:
Molecular Weight:
972.84
Beilstein:
79627
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (HPLC)

form

powder

optical activity

[α]20/D +136±3°, c = 10% in H2O

loss

≤10% loss on drying

mp

>278 °C (dec.) (lit.)

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C36H60O30/c37-1-7-25-13(43)19(49)31(55-7)62-26-8(2-38)57-33(21(51)15(26)45)64-28-10(4-40)59-35(23(53)17(28)47)66-30-12(6-42)60-36(24(54)18(30)48)65-29-11(5-41)58-34(22(52)16(29)46)63-27-9(3-39)56-32(61-25)20(50)14(27)44/h7-54H,1-6H2/t7-,8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-/m1/s1

InChI key

HFHDHCJBZVLPGP-RWMJIURBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Cyclodextrin (α-CD) is a water-soluble cyclic oligosaccharide that consists of six glucopyranose units linked by α-(1,4) bonds. It is commonly used to form inclusion complexes (ICs) with a wide range of organic compounds.

Application

α-Cyclodextrin may be used to synthesize a polymer inclusion complex (PIC) with poly(ethylene glycol) (PEG)-modified chitosans. These PICs can self-assemble to form supramolecular hydrogels.

Other Notes

Cyclodextrins are low molecular weight organic molecules which form watersoluble complexes with various compounds (guests); Enzyme model

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inclusion complexation between comblike PEO grafted polymers and ?-cyclodextrin.
He L
Macromolecules, 38(8), 3351-3355 (2005)
I. Tabushi
Tetrahedron, 40, 269-269 (1984)
Supramolecular Hydrogel Formation Based on Inclusion Complexation Between Poly (ethylene glycol)?Modified Chitosan and ??Cyclodextrin.
Huh K
Macromolecular Bioscience, 4(2), 92-99 (2004)
Taste modification of amino acids and protein hydrolysate by a-cyclodextrin
Linde GA, et al.
Food Research International, 42(7), 814-818 (2009)
M.L. Bender et al.
Cyclodextrin Chemistry (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service