Merck
  • β-Cyclodextrin's orientation onto TiO2 and its paradoxical role in guest's photodegradation.
β-Cyclodextrin's orientation onto TiO2 and its paradoxical role in guest's photodegradation.
Chemical communications (Cambridge, England) (2012-12-14)
Xu Zhang, Zixin Yang, Xuankun Li, Nansheng Deng, Shahua Qian
ABSTRACT
This work revealed that β-cyclodextrin was attached onto the surface of TiO(2) predominately by its secondary ring side, which caused paradoxical functions of β-cyclodextrin in the photodegradation of the four bisphenols. The equilibrium between the guest adsorbed through β-cyclodextrin onto TiO(2) and the one locked in β-CD in water could also change the role of β-cyclodextrin in the degradation of a certain guest.
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG, ≥98.0% (HPLC)
Sigma-Aldrich
β-Cyclodextrin, ≥97%
Sigma-Aldrich
β-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG, ≥95.0% (HPLC)