Skip to Content
Merck
All Photos(1)

Key Documents

76492

Sigma-Aldrich

10,12-Pentacosadiynoic acid

≥97% (HPLC)

Synonym(s):

PCDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H42O2
CAS Number:
Molecular Weight:
374.60
Beilstein:
4189973
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97% (HPLC)

form

crystals

functional group

carboxylic acid

SMILES string

CCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O

InChI

1S/C25H42O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25(26)27/h2-12,17-24H2,1H3,(H,26,27)

InChI key

ZPUDRBWHCWYMQS-UHFFFAOYSA-N

Application

10,12-Pentacosadiynoic acid (PCDA) is an amphiphilic diacetylene that can be used:
  • In combination with a semifluorinated tetracosane ((CF3(CF2)7(CH2)15CH3) to synthesize mixed monolayer by photopolymerization.
  • For the conversion of hydrophobic upconverting nanoparticles (UCNPs) to hydrophilic.
  • To synthesize a polymer by combining with pyridine-2-aldoxime for colorimetric detection of malathion.
  • To synthesize a thermochromic sensor constituting polymer fibres with monomeric unit of two PCDA linked by a p-phenylene group (bis-PCDA-Ph).
  • As an inert coating material upon self-assembly and photopolymerization to modify the surface of Fe3O4 nanoparticles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Seokho Kim et al.
Polymers, 12(3) (2020-03-14)
We experimentally demonstrate the simultaneous enhancement of Raman and photoluminescence (PL) of core-shell hybrid nanoparticles consisting of Ag (core) and polydiacetylene (PDA, shell) through the assistance of localized surface plasmon (LSP) effect for the effective biosensor. Core-shell nanoparticles (NPs) are
Thanh Chung Pham et al.
ACS omega, 5(48), 31254-31261 (2020-12-17)
We prepared the monomer PCDA-HP composed of 5-hydroxy-N 1,N 3-bis(pyridin-2-ylmethyl)isophthalamide (HP) as a cadmium ion tweezer and then polymerized them to form a polydiacetylene (PDA)-based sensor (PDA-HP), which displayed selective and sensitive colorimetric and fluorometric change upon addition of a
Ting Wang et al.
Nanoscale, 9(2), 869-874 (2016-12-21)
A flexible, transparent, and portable wrist strap sensor device has been well developed from a hierarchical polydiacetylene/MoS
One and a half layers? Mixed Langmuir monolayer of 10, 12-pentacosadiynoic acid and a semifluorinated tetracosane.
Wang S, et al.
Langmuir, 16(6), 2882-2886 (2000)
Chenlu He et al.
Nature communications, 11(1), 1188-1188 (2020-03-07)
Chiral noble metal nanoparticles has recently gained great interest due to their potential applications including ultrasensitive chiral recognition and asymmetric synthesis. We anticipate that they could be utilized to induce asymmetric photo-polymerization reactions with high enantioselectivity and reactivity. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service