764817
Poly(lactide-co-glycolide)-block-poly(ethylene glycol)-block-poly(lactide-co-glycolide)
average Mn (1100-1000-1100), lactide:glycolide 75:25
Synonym(s):
PLGA-PEG-PLGA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
description
typical PEG PDI<1.1; overall PDI≤2.0 (THF, PS)
Quality Level
form
semisolid
feed ratio
lactide:glycolide 75:25
mol wt
PEG average Mn 1,000
PLGA average Mn 2,200
average Mn (1100-1000-1100)
degradation timeframe
2-3 weeks
transition temp
Tm 230-235 °C
PDI
<2.0
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
PLGA-PEG-PLGA is an amphiphilic triblock copolymer which can self-assemble into micelles in aqueous medium due to the hydrophobic interactions present in the hydrophobic segments. The PEG segment imparts hydrophilicity and improves the biocompatibility of the copolymer. The PLGA segment forms a hydrophobic core and can solubilize hydrophobic drugs. These copolymers are widely used as nanocarriers for the sustained release of drugs.
Application
Used in the synthesis of targeted nanoparticles which are used for differential delivery and controlled release of drugs.
Features and Benefits
Biocompatible, degradable, thermosensitive, high stability, small size (<200 nm) and properties can be easily modified.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Zhimei Song et al.
Journal of colloid and interface science, 354(1), 116-123 (2010-11-04)
The aim of this study was to assess the potential of new copolymeric micelles to modify the pharmacokenetics and tissue distribution of Curcumin (CUR), a hydrophobic drug. In the present study, a poly (d,l-lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(d,l-lactide-co-glycolide) (PLGA-PEG-PLGA) copolymer was synthesized and
Frank Gu et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(7), 2586-2591 (2008-02-15)
There has been progressively heightened interest in the development of targeted nanoparticles (NPs) for differential delivery and controlled release of drugs. Despite nearly three decades of research, approaches to reproducibly formulate targeted NPs with the optimal biophysicochemical properties have remained
PLGA-PEG Encapsulated sitamaquine nanoparticles drug delivery system against Leishmania donovani
Kumara, R., Sahoo, G. C., Pandeya, K., Dasa, V. N. R., Yousuf, M., Ansaria, S. R., & Dasa, P.
Journal of Scientific and Innovative Research, 3(1), 85-90 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service