Skip to Content
Merck
All Photos(1)

Key Documents

764779

Sigma-Aldrich

Poly(ethylene glycol) methyl ether-block-poly(D,L lactide)

PEG average Mn 2,000, PDLLA average Mn 2,000

Synonym(s):

PEG-PDLLA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

description

n=45, m=28 (typical)
typical PEG PDI<1.1; overall PDI<1.4 (THF, PEO)

form

pellets

mol wt

PDLLA average Mn 2,000
PEG average Mn 2,000
average Mn 4,000 (total)

degradation timeframe

2-4 weeks

transition temp

Tg 11 °C (PEG block)
Tm 244-248 °C
Tg 88 °C (PDLLA block)

PDI

≤1.4

storage temp.

2-8°C

General description

Block copolymer micelles are widely used in drug delivery applications. PEG-PDLLA is a biodegradable polymeric micelle which is used as carriers for hydrophobic drugs like paclitaxel. PEG is the hydrophilic part and PDLLA is the hydrophobic part which forms the micelle core wherein the hydrophobic drug is loaded.

Application

Used as a carrier for the controlled and targeted release of anticancer hydrophobic drugs.
water-solubilization of drugs

Features and Benefits

  • Good biocompatibility, low immunogenicity and good degradability.
  • Properties can be easily modulated by changing the block copolymer segment sizes to suit a particular application.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hongtao Chen et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(18), 6596-6601 (2008-05-01)
It is generally assumed that polymeric micelles, upon administration into the blood stream, carry drug molecules until they are taken up into cells followed by intracellular release. The current work revisits this conventional wisdom. The study using dual-labeled micelles containing
Arunvel Kailasan et al.
Acta biomaterialia, 6(3), 1131-1139 (2009-09-01)
This work describes the synthesis and characterization of novel thermoresponsive highly branched polyamidoamine-polyethylene glycol-poly(D,L-lactide) (PAMAM-PEG-PDLLA) core-shell nanoparticles. A series of dendritic PEG-PDLLA nanoparticles were synthesized through conjugation of PEG of various chain lengths (1500, 6000 and 12,000 g mol(-1)) to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service