Skip to Content
Merck
All Photos(1)

Key Documents

914460

Sigma-Aldrich

1-Biotinyl-3,6-dioxa-8-octaneamine hydrochloride

≥95%

Synonym(s):

1-Biotinyl-3,6-dioxa-8-octaneamine hydrochloride, N-(2-(2-(2-aminoethoxy)ethoxy)ethyl)-5-(2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide hydrochloride, N-{2-[2-(2- Aminoethoxy)ethoxy]ethoxy}biotinamide hydrochloride, Amine-terminated biotin linker, Biotin-DOOA HCl, Biotinylation reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H31ClN4O4S
CAS Number:
Molecular Weight:
410.96
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

storage temp.

2-8°C

SMILES string

S1[C@H]([C@H]2NC(=O)N[C@H]2C1)CCCCC(=O)NCCOCCOCCN.Cl

InChI

1S/C16H30N4O4S.ClH/c17-5-7-23-9-10-24-8-6-18-14(21)4-2-1-3-13-15-12(11-25-13)19-16(22)20-15;/h12-13,15H,1-11,17H2,(H,18,21)(H2,19,20,22);1H/t12-,13-,15-;/m0./s1

InChI key

UNQYTGLJZONNBD-HZPCBCDKSA-N

Related Categories

Application

1-Biotinyl-3,6-dioxa-8-octaneamine hydrochloride is a versatile biotinylated linker that can be incorporated into chemical tools via its terminal amino group. Labeling materials or proteins with biotin provides a means to enrich and capture targets from biological systems.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiyuan Fang et al.
Chemical communications (Cambridge, England), 49(55), 6164-6166 (2013-06-04)
A direct ELISA was established for the fast detection of semicarbazide (SEM) using a novel biotin derivative. Without a tedious extraction procedure, as low as 0.07 μg L(-1) of SEM could be detected reproducibly. This assay has better recovery and
Wilke C de Vries et al.
ACS applied materials & interfaces, 9(48), 41760-41766 (2017-11-16)
We present the preparation of ligand-conjugated redox-responsive polymer nanocontainers by the supramolecular decoration of cyclodextrin vesicles with a thin redox-cleavable polymer shell that displays molecular recognition units on its surface. Two widely different recognition motifs (mannose-Concanavalin A and biotin-streptavidin) are
Dilini N Kekulandara et al.
Biochemistry, 57(5), 772-780 (2017-12-21)
Thioredoxin 1 (Trx1) and glutaredoxin 1 (Grx1) are two ubiquitous redox enzymes that are central for redox homeostasis but also are implicated in many other processes, including stress sensing, inflammation, and apoptosis. In addition to their enzymatic redox activity, a
Sebastian Pomplun et al.
Angewandte Chemie (International ed. in English), 58(11), 3542-3547 (2019-01-18)
We discovered N-pyrrolyl alanine derivatives as efficient reagents for the fast and selective Pictet-Spengler reaction with aldehyde-containing biomolecules. Other aldehyde-labeling methods described so far have several drawbacks, like hydrolytic instability, slow reaction kinetics or not readily available labeling reagents. Pictet-Spengler
Yali Yuan et al.
Chemical communications (Cambridge, England), 54(80), 11352-11355 (2018-09-25)
We have developed highly fluorescent, monolithic colloidal CdSe seeded CdS nanorod clusters comprising thousands of nanorods. Their use in the sandwich assay detection of a model protein yields a thousand-fold improvement in the detection limit compared to individual nanorods, making

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service