Skip to Content
Merck
All Photos(1)

Key Documents

763543

Sigma-Aldrich

1,1,1-Tris[(dodecylthiocarbonothioylthio)-2-methylpropionate]ethane

98% (HPLC)

Synonym(s):

3 arm star polymer RAFT agent, Tris(DDMAT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H102O6S9
Molecular Weight:
1159.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Assay

98% (HPLC)

form

liquid

refractive index

n20/D 1.5314

density

1.085 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCSC(=S)SC(C)(C)C(=O)OCC(C)(COC(=O)C(C)(C)SC(=S)SCCCCCCCCCCCC)COC(=O)C(C)(C)SC(=S)SCCCCCCCCCCCC

InChI

1S/C56H102O6S9/c1-11-14-17-20-23-26-29-32-35-38-41-66-50(63)69-53(4,5)47(57)60-44-56(10,45-61-48(58)54(6,7)70-51(64)67-42-39-36-33-30-27-24-21-18-15-12-2)46-62-49(59)55(8,9)71-52(65)68-43-40-37-34-31-28-25-22-19-16-13-3/h11-46H2,1-10H3

InChI key

HYZIWJXWRIAPNA-UHFFFAOYSA-N

General description

Functionalized RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene; acrylate and acrylamide monomers. For use in advanced material (rheology; etc) and biomaterial (nanomedicine) applications. Chain Transfer Agent (CTA)
Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

Reversible Addition Fragmentation Chain Transfer (RAFT) Polymerization

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service