761621
Fluor 488-Alkyne
95% (HPLC)
Synonym(s):
Rhodamine 110-PEG4-alkyne
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C32H33N3O8
Molecular Weight:
587.62
UNSPSC Code:
12352200
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95% (HPLC)
form
solid
reaction suitability
reaction type: click chemistry
storage temp.
−20°C
Application
Rhodamine 110 conjugated terminal alkyne. Can be used to detect or label azide containing molecules or biomolecules by fluorescence spectroscopy following copper catalyzed azide-alkyne cycloaddition.
Spectral Properties: Abs/Em = 501/525

Spectral Properties: Abs/Em = 501/525

Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service