757853
Poly(ethylene glycol) (N-hydroxysuccinimide 5-pentanoate) ether N′-(3-maleimidopropionyl)aminoethane
average MN 4,000, cross-linking reagent thiol reactive, cross-linking reagent amine reactive, NHS ester, maleimide
Synonym(s):
Maleimide polyoxyethylene NHS-protected valeric acid, Maleimide-PEG-NHS protected acid, Maleimide-PEG-acid, Polyethylene glycol, maleimide-PEG-valeric acid
About This Item
Recommended Products
product name
Poly(ethylene glycol) (N-hydroxysuccinimide 5-pentanoate) ether N′-(3-maleimidopropionyl)aminoethane, average Mn 4,000
form
solid
Quality Level
mol wt
average Mn 4,000
reaction suitability
reagent type: cross-linking reagent
reactivity: amine reactive
reagent type: cross-linking reagent
reactivity: thiol reactive
transition temp
Tm 46-51 °C
Ω-end
NHS ester
α-end
maleimide
polymer architecture
shape: linear
functionality: heterobifunctional
storage temp.
−20°C
Related Categories
General description
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service