Skip to Content
Merck
All Photos(1)

Documents

713783

Sigma-Aldrich

O,O′-Bis[2-(N-Succinimidyl-succinylamino)ethyl]polyethylene glycol

2,000

Synonym(s):

Polyethylene glycol, α,ω-Bis-NHS-PEG, Di(N-succinimidyl) PEG-diacid, PEG-bis(N-succinimidyl succinate)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.22

form

powder

mol wt

average Mn 2,000

reaction suitability

reagent type: cross-linking reagent

Ω-end

NHS ester

α-end

NHS ester

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

O,O′-Bis[2-(N-Succinimidyl-succinylamino)ethyl]polyethylene glycol (NHS-PEG-NHS) is a cross-linking reagent.

Application

O,O′-Bis[2-(N-Succinimidyl-succinylamino)ethyl]polyethylene glycol (NHS-PEG-NHS) is suitable reagent used in the covalent conjugation of peanut agglutinin and anticarcinoembryonic antigen antibodies (αCEA) tumor-targeting ligands to near-infrared (NIR)fluorescent superparamagnetic iron oxide (IO)- human serum albumin (HSA) nanoparticles. It is suitable reagent used in the synthesis of novel series of mesoporous silica nanoparticles with different pore sizes, useful for high loading and controlled release of short oligonucleotides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jixi Zhang et al.
Dalton transactions (Cambridge, England : 2003), 43(10), 4115-4126 (2014-01-25)
Aimed at high loading and controlled release of oligonucleotides with short sequences of base-pairs, a novel series of mesoporous silica nanoparticles with three different pore sizes (3.5-5.0 nm) but the same cleavable surface linkers (MSN-Linker-Cys) were synthesized. The small particle
Enav Corem-Salkmon et al.
International journal of nanomedicine, 7, 5517-5527 (2012-11-01)
Colon cancer is one of the major causes of death in the Western world. Early detection significantly improves long-term survival for patients with the disease. Near- infrared (NIR) fluorescent nanoparticles hold great promise as contrast agents for tumor detection. NIR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service