Skip to Content
Merck
All Photos(1)

Documents

755850

Sigma-Aldrich

11-Maleimidoundecanoic acid

95% (GC)

Synonym(s):

11-Maleimide undecanoic acid, 2,5-Dihydro-2,5-dioxo-1H-pyrrole-1-undecanoic acid, MM-281, MUDA, Maleimidoundecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95% (GC)

form

powder

mp

94-98 °C

SMILES string

OC(=O)CCCCCCCCCCN1C(=O)C=CC1=O

InChI

1S/C15H23NO4/c17-13-10-11-14(18)16(13)12-8-6-4-2-1-3-5-7-9-15(19)20/h10-11H,1-9,12H2,(H,19,20)

InChI key

UVZTZBRGZXIBLZ-UHFFFAOYSA-N

General description

Intermediate for ester and amide linked maleimide monomers, can be used to generate liquid crystal copolymers

Application

The maleimide functional group can be used to conjugate a variety of biomolecules such as enzymes and DNA to the polymer chain.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safak, M.; Alemdaroglu, F. E.; Li, Y.; Ergen, E.; Hernnann, A.
Advanced Materials, 19, 1499-1499 (2007)
Pietro Buono et al.
ChemSusChem, 10(5), 984-992 (2017-01-04)
In the present report an environmentally friendly approach to transforming renewable feedstocks into value-added materials is proposed. This transformation pathway was conducted under green conditions, without the use of solvents or catalyst. First, controlled modification of lignin, a major biopolymer
Karina L Heredia et al.
Macromolecules, 42(7), 2360-2367 (2009-01-01)
In this report we describe a straightforward approach to synthesize polymers with end-groups that bind site-specifically to two different proteins. Telechelic biotin, maleimide poly(N-isopropylacrylamide) (pNIPAAm) was synthesized for the formation of streptavidin (SAv)-bovine serum albumin (BSA) polymer conjugates. Reversible addition-fragmentation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service