Skip to Content
Merck
All Photos(4)

Key Documents

753009

Sigma-Aldrich

SPhos Pd G2

Synonym(s):

2nd Generation SPhos Precatalyst, Chloro(2-dicyclohexylphosphino-2′,6′-dimethoxy-1,1′-biphenyl)[2-(2′-amino-1,1′-biphenyl)]palladium(II), SPhos-Pd-G2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H45ClNO2PPd
CAS Number:
Molecular Weight:
720.62
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

feature

generation 2

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

218 °C (dec.)

functional group

phosphine

SMILES string

COC1=CC=CC(OC)=C1C2=C(P(C3CCCCC3)C4CCCCC4)C=CC=C2.NC5=C(C6=C([Pd]Cl)C=CC=C6)C=CC=C5

InChI

1S/C26H35O2P.C12H10N.ClH.Pd/c1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h9-11,16-21H,3-8,12-15H2,1-2H3;1-6,8-9H,13H2;1H;/q;;;+1/p-1

InChI key

NOMWEFBAPVZIIP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

SPhos Pd G2 is a second generation (G2) precatalyst in which phenethylamine based backbone of the (G1) complex is substituted with a biphenyl-based ligand. This improvement on the previous generation allows creating the active palladium species at room temperature with weak phosphate or carbonate bases. The catalyst proves remarkably adept at achieving a variety of Suzuki-Miyaura couplings among other cross-coupling reactions.

Application

Application in Suzuki-Miyaura coupling:
  • Self-arylation of 1-alkyl-3-bromo-2-phenyl-2,1-borazaronaphthalenes to form 2,1-borazaronaphthols.
  • Suzuki–Miyaura coupling of [Ru(bpy)2]+ with 4-carboxyphenylboronic acid.
  • Suzuki–Miyaura coupling of β-borylated porphyrins with 2-iodoaniline.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modular synthesis of simple cycloruthenated complexes with state-of-the-art performance in p-type DSCs
Brunner F, et al.
Journal of Material Chemistry C, 4(41), 9823-9833 (2016)
Pictet-Spengler Synthesis of Quinoline-Fused Porphyrins and Phenanthroline-Fused Diporphyrins
Gao K, et al.
Angewandte Chemie (International Edition in English), 55(42), 13038-13042 (2016)
Accessing 2, 1-Borazaronaphthols: Self-Arylation of 1-Alkyl-2-aryl-3-bromo-2, 1-borazaronaphthalenes
Molander, Gary A and Wisniewski, Steven R
The Journal of Organic Chemistry, 79(17), 8339-8347 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service