Skip to Content
Merck
All Photos(1)

Documents

756741

Sigma-Aldrich

PCy3 Pd G2

95%

Synonym(s):

Chloro[(tricyclohexylphosphine)-2-(2′-aminobiphenyl)]palladium(II), Tricyclohexylphosphine Pd G2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H43ClNPPd
CAS Number:
Molecular Weight:
590.52
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

feature

generation 2

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: C-H Activation
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

244-246 °C

functional group

phosphine

SMILES string

NC1=C(C2=C([Pd]Cl)C=CC=C2)C=CC=C1.C3(CCCCC3)P(C4CCCCC4)C5CCCCC5

InChI

1S/C18H33P.C12H10N.ClH.Pd/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h16-18H,1-15H2;1-6,8-9H,13H2;1H;/q;;;+1/p-1

InChI key

CYJRABDADOSMKA-UHFFFAOYSA-M

Application

PCy3 Pd G2 is a monophosphine-coordinated 2-phenylaniline-based palladacycle complex that can be used as a precatalyst:
  • To prepare diarylmethanes by Suzuki cross-coupling reaction with heterocyclic-chloromethyl derivatives with aryl/heteroaryl boronic acids.
  • To synthesize poly(arylene)s by the Suzuki cross-coupling polymerization reaction between aryl dihalides and aryldiboronic acids.
  • In the C-H bond functionalization reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

t-Bu3P-coordinated 2-phenylaniline-based palladacycle complex as a precatalyst for the Suzuki cross-coupling polymerization of aryl dibromides with aryldiboronic acids
Zhang Hong-Hai, et al.
ACS Macro Letters, 2, 10-13 (2013)
Facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes
Schmink JR and Tudge MT
Tetrahedron Letters, 54, 15-20 (2013)
Serge I Gorelsky et al.
The Journal of organic chemistry, 77(1), 658-668 (2011-12-14)
A comprehensive understanding of the C-H bond cleavage step by the concerted metalation-deprotonation (CMD) pathway is important in further development of cross-coupling reactions using different catalysts. Distortion-interaction analysis of the C-H bond cleavage over a wide range of (hetero)aromatics has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service