Skip to Content
Merck
All Photos(7)

Key Documents

638072

Sigma-Aldrich

SPhos

greener alternative

98%

Synonym(s):

SPhos, 2-Dicyclohexylphosphino-2′,6′-dimethoxybiphenyl, S Phos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H35O2P
CAS Number:
Molecular Weight:
410.53
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Kumada Coupling

reagent type: ligand
reaction type: Negishi Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

greener alternative product score

old score: 10
new score: 1
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Less Hazardous Chemical Syntheses
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

164-166 °C (lit.)

functional group

phosphine

greener alternative category

SMILES string

COc1cccc(OC)c1-c2ccccc2P(C3CCCCC3)C4CCCCC4

InChI

1S/C26H35O2P/c1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21/h9-11,16-21H,3-8,12-15H2,1-2H3

InChI key

VNFWTIYUKDMAOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SPhos [2-Dicyclohexylphosphino-2′,6′-dimethoxybiphenyl] is an air-stable, electron-rich biaryl phosphine ligand developed by the Buchwald group to enhance the reactivity of palladium catalysis during cross-coupling reactions.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Less Hazardous Chemical Syntheses” and “Inherently Safer Chemistry for Accident Prevention”. Click here to view its DOZN scorecard.

Application

Learn more about Buchwald Phosphine Ligands

For small scale and high throughput uses, product is also available as ChemBeads (932191)
Highly universal ligand for Suzuki-Miyaura coupling; aryl chlorides, hindered biaryls, heterobiaryls.
SPhos may be used as a ligand in the following processes:
  • Palladium catalyzed Suzuki-Miyaura cross-coupling reaction between Boc-protected aminomethyltrifluoroborate and aryl chlorides or hetaryl chlorides to form the corresponding aminomethylarenes.[1]
  • Palladium catalyzed Suzuki-Miyaura cross-coupling reaction between 4-methyl-substituted piperidinylzinc reagent and different aryl or heteroaryl iodides to form various substituted piperidines.[2]
  • Intramolecular Suzuki-Miyaura coupling to form the 18-membered macrocyclic ring during the multi-step synthesis of riccardin C.[3]
Utilized in conjunction with palladium to form a highly active catalyst for C-N bond formation.[4]

Legal Information

Usage subject to Patents: US 6307087; EP 1097158; JP 5758844; CA 2336691

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of the SPhos Ligand to Suppress Racemization in Arylpinacolboronate Ester Suzuki Couplings Involving ?-Amino Acids. Synthesis of Biaryl Derivatives of 4-Hydroxyphenylglycine, Tyrosine, and Tryptophan
Prieto, Monica, et al.
The Journal of Organic Chemistry, 74.23, 9202-9205 (2009)
Highly diastereoselective arylations of substituted piperidines
Seel, Stephanie, et al.
Journal of the American Chemical Society, 133.13, 4774-4777 (2011)
What makes for a good catalytic cycle? A theoretical study of the SPhos ligand in the Suzuki?Miyaura reaction
Kozuch, Sebastian, and Jan ML Martin
Chemical Communications (Cambridge, England), 47.17, 4935-4937 (2011)
Synthesis of 1, 5-disubstituted tetrazoles via Suzuki?Miyaura cross-coupling of 5-chloro-1-phenyltetrazole
Tang, Qing, and Ryan Gianatassio
Tetrahedron Letters, 51.27, 3473-3476 (2010)
Efficient synthesis of functionalized dibenzofurans by domino `twofold Heck/6?-electrocyclization?reactions of 2, 3-di-and 2, 3, 5-tribromobenzofuran
Hussain, Munawar, Nguyen Thai Hung, and Peter Langer
Tetrahedron Letters, 50.27, 3929-3932 (2009)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service