Skip to Content
Merck
All Photos(1)

Key Documents

752126

Sigma-Aldrich

Ethylmagnesium bromide solution

40% in 2-methyltetrahydrofuran

Synonym(s):

Bromoethylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2MgBr
CAS Number:
Molecular Weight:
133.27
Beilstein:
3587203
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

40% in 2-methyltetrahydrofuran

density

1.140 g/mL at 25 °C

SMILES string

CC[Mg]Br

InChI

1S/C2H5.BrH.Mg/c1-2;;/h1H2,2H3;1H;/q;;+1/p-1

InChI key

TWTWFMUQSOFTRN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janne E Tønder et al.
Organic & biomolecular chemistry, 2(10), 1447-1455 (2004-05-12)
Copper mediated allylic substitutions and conjugate additions to geranyl, cinnamyl and allylic indole compounds have been investigated with the aim of finding a method for the creation of the all-carbon quaternary centre present in the natural product lyngbyatoxin A. Reaction
T Nishiyama et al.
Chemical & pharmaceutical bulletin, 48(12), 1999-2002 (2001-01-06)
The direct alpha-bromination of various ketones using trifluoromethanesulfonic anhydride and Grignard reagent or magnesium bromide in ether gave the corresponding alpha-bromo ketones in moderate to good yields under mild reaction conditions.
F Turon et al.
Lipids, 37(8), 817-821 (2002-10-10)
An analytical procedure was developed for regiodistribution analysis of TAG using alpha-MAG prepared by an ethyl magnesium bromide deacylation. In the present communication, the deacylation procedure is shown to lead to representative alpha-MAG, allowing the composition of the native TAG
R G Xie et al.
Steroids, 55(11), 488-490 (1990-11-01)
The reaction of ethyl magnesium bromide and 17 alpha-ethynylestradiol with formaldehyde in the presence of triethyl phosphate or hexamethylphosphoramide gave the 2- and 4-formyl-17 alpha-ethynylestradiol in high yield. Treatment of the formyl derivatives with an alkaline solution of hydrogen peroxide
Yasuhiro Ando et al.
Journal of oleo science, 57(8), 459-462 (2008-07-16)
This paper presents a procedure for preparation of a Grignard reagent, ethyl magnesium bromide, used for partial deacylation of triacylglycerols (TAG) in their regiospecific analysis. Magnesium turnings were reacted with ethereal solution of bromoethane in a screw-capped test tube to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service