All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C11H20N2
CAS Number:
Molecular Weight:
180.29
Beilstein:
122643
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98.0%
form
liquid
refractive index
n20/D 1.475
density
0.91 g/mL at 20 °C (lit.)
SMILES string
CCCCCCCCn1ccnc1
InChI
1S/C11H20N2/c1-2-3-4-5-6-7-9-13-10-8-12-11-13/h8,10-11H,2-7,9H2,1H3
InChI key
KLMZKZJCMDOKFE-UHFFFAOYSA-N
Other Notes
Ligand employed in the highly enantioselective epoxidation of olefins by Jacobsen′s catalyst
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J J Yourick et al.
Biochemical pharmacology, 42(7), 1361-1366 (1991-09-12)
In humans and animals, disulfiram produces a disulfiram-ethanol reaction after an ethanol challenge, the basis of which is the inhibition of liver aldehyde dehydrogenase (ALDH). Disulfiram and the metabolites diethyldithiocarbamate (DDTC), diethyldithiocarbamate-methyl ester (DDTC-Me), and S-methyl-N,N-diethylthiolcarbamate (DETC-Me) were studied in
K. Imagawa et al.
Chemistry Letters (Jpn), 527-527 (1994)
Mounesha N Garaga et al.
Nanoscale, 10(26), 12337-12348 (2018-05-22)
We report enhanced protonic and ionic dynamics in an imidazole/protic ionic liquid mixture confined within the nanopores of silica particles. The ionic liquid is 1-octylimidazolium bis(trifluoromethanesulfonyl)imide ([HC8Im][TFSI]), while the silica particles are microsized and characterized by internal well connected nanopores.
V Ravindranath et al.
Toxicology and applied pharmacology, 78(3), 370-376 (1985-05-01)
2-Methylfuran (2-MF), a constituent of cigarette smoke and coffee, causes necrosis of liver, lungs, and kidneys in rodents. 2-MF is metabolically activated by mixed-function oxidases to acetylacrolein, a reactive metabolite that binds covalently to microsomal protein. The hepatic microsomal metabolism
Jonas Winters et al.
Physical chemistry chemical physics : PCCP, 21(7), 4053-4062 (2019-02-05)
Polyaramids are a class of high-performance polymers, known for their high mechanical strength and chemical and thermal stability. Their ability to create a network of intermolecular hydrogen bonds causes them to be very poorly soluble in conventional solvents. Hazardous solvents
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service