Skip to Content
Merck
All Photos(4)

Documents

136131

Sigma-Aldrich

1,2-Dimethylimidazole

97%

Synonym(s):

1,2-Dimethyl-1H-imidazole, 1,2DMZ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N2
CAS Number:
Molecular Weight:
96.13
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 20 °C)

Assay

97%

form

solid

autoignition temp.

896 °F

bp

204 °C (lit.)

mp

37-39 °C (lit.)

density

1.084 g/mL at 25 °C (lit.)

SMILES string

Cc1nccn1C

InChI

1S/C5H8N2/c1-5-6-3-4-7(5)2/h3-4H,1-2H3

InChI key

GIWQSPITLQVMSG-UHFFFAOYSA-N

General description

1,2-Dimethylimidazole reacts with copper(II) tetraacetate to yield cis-bis(acetato)bis(1,2-dimethylimidazole) copper(II) complex. It undergoes Menschutkin reaction with benzyl bromide to yield 3-benzyl-1,2-dimethylimidazolium bromide and kinetics of reaction was studied in various ionic liquids and molecular organic solvents.

Application

1,2-Dimethylimidazole was used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It was also used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wesley A Henderson et al.
Chemical communications (Cambridge, England), (35)(35), 3708-3710 (2006-10-19)
The crystal structures and thermal behavior of the 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexa-fluorophosphate salts are compared with the analogous 1-butyl-2,3-dimethylimidazolium salts to examine the influence of the ether oxygen on salt thermal properties for a typical constituent cation used in the
The coordination environment of mitochondrial cytochromes b.
K R Carter et al.
FEBS letters, 132(2), 243-246 (1981-09-28)
Physical properties of substituted imidazolium based ionic liquids gel electrolytes.
Sutto TE, et al.
Zeitschrift fur Naturforschung, A, 57(11), 839-846 (2002)
M M Fitzgerald et al.
Protein science : a publication of the Protein Society, 4(9), 1844-1850 (1995-09-01)
The activated state of cytochrome c peroxidase, compound ES, contains a cation radical on the Trp-191 side chain. We recently reported that replacing this tryptophan with glycine creates a buried cavity at the active site that contains ordered solvent and
Alain Lesimple et al.
Journal of mass spectrometry : JMS, 43(1), 35-41 (2007-07-10)
A number of 1,2-dimethylimidazole ionic liquids substituted on N(II) with alkyl chains of varying lengths terminated with sulfur-containing groups were investigated by electrospray high-resolution tandem Fourier-transform mass spectrometry. Fragmentation pathways are strongly dependent on the oxidation state of the sulfur

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service