750492
Carbon nanotube, single-walled
<3.5% Metal Catalyst, avg. no. of layers, 1
Synonym(s):
PureTubes™, SWCNT, SWNT, Single wall carbon nanotube
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
30% (Metallic)
70% (Semiconducting)
Quality Level
form
solid
feature
avg. no. of layers 1
manufacturer/tradename
NanoIntegris, Inc.
L
0.3-5 μm
impurities
<3.5% Metal Catalyst
diameter
1.2-1.7 nm
mp
3652-3697 °C (lit.)
density
1.7-1.9 g/cm3 at 25 °C (lit.)
SMILES string
[C]
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
PureTubes™ are produced by arch discharge or plasma torch. The diameter of the semiconducting single walled nanotubes range in 1.2-1.7 nm, whereas the length varies from 100nm to 4 μm. The catalyst content is < 3.5%.
Application
These carbon nanotubes are produced via the Arc Discharge process and are purified via density gradient centrifugation.
Research applications include:
Research applications include:
- Electronics
- Sensors
- Composites
- Energy Storage
- Study of Life Science systems
Preparation Note
Electric Arc Discharge Method
Legal Information
Product of NanoIntegris, Inc
PureTubes is a trademark of NanoIntegris, Inc.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Skin Sens. 1 - STOT RE 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chad D Vecitis et al.
ACS nano, 4(9), 5471-5479 (2010-09-04)
Single-walled carbon nanotubes (SWNTs) have been previously observed to be strong antimicrobial agents, and SWNT coatings can significantly reduce biofilm formation. However, the SWNT antimicrobial mechanism is not fully understood. Previous studies on SWNT cytotoxicity have concluded that membrane stress
Chuan Wang et al.
ACS nano, 4(12), 7123-7132 (2010-11-11)
Macroelectronic integrated circuits are widely used in applications such as flat panel display and transparent electronics, as well as flexible and stretchable electronics. However, the challenge is to find the channel material that can simultaneously offer low temperature processing, high
Amin Salehi-Khojin et al.
ACS nano, 5(1), 153-158 (2010-12-29)
There has been recent controversy whether the response seen in carbon nanotube (CNT) chemiresistors is associated with a change in the resistance of the individual nanotubes or changes in the resistance of the junctions. In this study, we carry out
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service