738573
Platinum-ruthenium alloy on graphitized carbon
extent of labeling: 20 wt. % Pt loading, extent of labeling: 10 wt. % Ru loading
Synonym(s):
20% Pt, 10% Ru on Vulcan XC72, 30% Pt-Ru/Vulcan
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
Pt/Ru
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
powder
Quality Level
reaction suitability
reagent type: catalyst
core: platinum
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
extent of labeling
20 wt. % Pt loading
10 wt. % Ru loading
particle size
<5 nm (Pt)
greener alternative category
SMILES string
[Ru].[Pt]
InChI
1S/Pt.Ru
InChI key
CFQCIHVMOFOCGH-UHFFFAOYSA-N
Application
Platinum-ruthenium alloy on graphitized carbon (30% Pt-Ru/Vulcan) can be used as an electrocatalyst for the fabrication of direct methanol fuel cells (DMFC) and proton exchange membrane (PEM) based fuel cells.
Other Notes
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
nwg
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Pt-WO3 supported on carbon nanotubes as possible anodes for direct methanol fuel cells
Rajesh B, et al.
Fuel: The Science and Technology of Fuel and Energy, 81(17), 2177-2190 (2002)
Zhaolin Liu et al.
Langmuir : the ACS journal of surfaces and colloids, 20(1), 181-187 (2005-03-05)
PtRu nanoparticles supported on Vulcan XC-72 carbon and carbon nanotubes were prepared by a microwave-assisted polyol process. The catalysts were characterized by transmission electron microscopy, X-ray diffraction, and X-ray photoelectron spectroscopy (XPS). The PtRu nanoparticles, which were uniformly dispersed on
Liu, L.; Pu, C.; Viswanathan, R.; Fan, Q.; Liu, R.; Smotkin, E. S.
Electrochimica Acta, 43, 3657-3657 (1998)
Pt-Ru electrocatalysts supported on ordered mesoporous carbon for direct methanol fuel cell
Salgado JRC, et al.
Journal of Power Sources, 195(13), 4022-4029 (2010)
CO-tolerance of low-loaded Pt/Ru anodes for PEM fuel cells
Qi Z and Kaufman A
Journal of Power Sources, 113(1), 115-123 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service