738557
Platinum on graphitized carbon
extent of labeling: 40 wt. % loading
Synonym(s):
40% Pt on Vulcan XC72, 40% Pt/Vulcan
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
powder
reaction suitability
reagent type: catalyst
core: platinum
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
extent of labeling
40 wt. % loading
particle size
<5 nm (Pt)
greener alternative category
SMILES string
[Pt]
InChI
1S/Pt
InChI key
BASFCYQUMIYNBI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Platinum on graphitized carbon (40% Pt/Vulcan XC72) is a high surface area material that is used as an electrocatalyst for the development of electrolytic fuel cells and direct methanol fuel cells. It can also be used in the formation of a non-enzymatic sensor for the oxidation of glucose.
Other Notes
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
nwg
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Development of Cu2O/Carbon Vulcan XC-72 as non-enzymatic sensor for glucose determination
El Khatib KM and Hameed RMA
Biosensors And Bioelectronics, 26(8), 3542-3548 (2011)
Load cycle durability of a graphitized carbon black-supported platinum catalyst in polymer electrolyte fuel cell cathodes
Takei C, et al.
Journal of Power Sources, 324, 729-737 (2016)
Oxygen reduction on a high-surface area Pt/Vulcan carbon catalyst: a thin-film rotating ring-disk electrode study
Paulus UA, et al.
Journal of Electroanalytical Chemistry, 495(2), 134-145 (2001)
Platinum nanoparticles supported by a Vulcan XC-72 and PANI doped with trifluoromethane sulfonic acid substrate as a new electrocatalyst for direct methanol fuel cells
Gharibi H, et al.
The Journal of Physical Chemistry C, 114(11), 5233-5240 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service