Skip to Content
Merck
All Photos(4)

Key Documents

206180

Sigma-Aldrich

Ruthenium on carbon

greener alternative

extent of labeling: 5 wt. % loading

Synonym(s):

Ruthenium on activated carbon, heterogeneous catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ru
Molecular Weight:
101.07
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

form

powder

reaction suitability

reagent type: catalyst
core: ruthenium

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

5 wt. % loading

matrix

carbon support

greener alternative category

SMILES string

[Ru]

InChI

1S/Ru

InChI key

KJTLSVCANCCWHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ruthenium on carbon is a powdered form of catalytic metal ruthenium supported on high-surface area conductive carbon, which makes it useful as an electrocatalyst. Ruthenium-based electrocatalysts are studied in fuel cells and water electrolyzers. It is also used in chemical hydrogenation reactions for example of the hydrogenation of aldehydes, ketones, sugars, and some aromatics
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Ruthenium on carbon (Ru/C) can be used as a heterogeneous noble metal catalyst for the hydrotreatment of fast pyrolysis of oil derived from lignocellulosic biomass. It can be used in combination with an Amberlyst heat resistant ion-exchange resin for the hydrogenolysis of glycerol to 1,2-propanediol.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-heat. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cong Wang et al.
JACS Au, 1(9), 1422-1434 (2021-10-05)
Plastics waste has become a major environmental threat, with polyethylene being one of the most produced and hardest to recycle plastics. Hydrogenolysis is potentially the most viable catalytic technology for recycling. Ruthenium (Ru) is one of the most active hydrogenolysis
Glycerol hydrogenolysis to 1, 2-propanediol catalyzed by a heat-resistant ion-exchange resin combined with Ru/C
Miyazawa T, et al.
Applied Catalysis A: General, 329(23), 30-35 (2007)
Hydrotreatment of fast pyrolysis oil using heterogeneous noble-metal catalysts
Wildschut J, et al.
Industrial & Engineering Chemistry Research, 48(23), 10324-10334 (2009)
Svilen P Simeonov et al.
ChemSusChem, 12(12), 2748-2754 (2019-05-06)
A new scalable synthesis of pentane-1,2,5-triol from the furanics platform has been developed. Excellent yields of up to 92 % are obtained under flow conditions by using readily available catalysts from the existing pool. The strategy exploits the highly functionalized Achmatowicz
T C Ooi et al.
Clinica chimica acta; international journal of clinical chemistry, 446, 97-104 (2015-04-11)
Circulating PCSK9 levels are higher in women than men, in postmenopausal than premenopausal women, and in pregnant than non-pregnant women, suggesting that sex hormones may be related to PCSK9 levels. We have examined the relationship between serum estradiol (E2) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service