Skip to Content
Merck
All Photos(5)

Key Documents

205915

Sigma-Aldrich

Platinum black

greener alternative

black, powder, ≤20 μm, ≥99.95% trace metals basis

Synonym(s):

Platinum gray, Pt, Pt Black

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.95% trace metals basis

form

powder

reaction suitability

core: platinum
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

~25 m2/g

particle size

≤20 μm

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Platinum black (black, powder, ≤20 μm) can be electroplated on implantable microelectrode arrays (MEAs) to improve the impedence of the device. It can also be used as a microelectrode for the formation of a microfluidic electrochemical sensor for glucose sensing applications. It can be used as a photocathode for the production of hydrogen.
Platinum black can be used as a catalyst to facilitate the electrochemical reactions in proton exchange membrane (PEM) fuel cells. These fuel cells are promising candidates as a clean and energy efficient power source.

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Composition and performance modelling of catalyst layer in a proton exchange membrane fuel cell
Marr C
Journal of Power Sources, 77(1), 17-27 (1999)
Optimum Nafion content in PEM fuel cell electrodes
Sasikumar G
Electrochimica Acta, 50.2-3, 601-605 (2004)
Improving impedance of implantable microwire multi-electrode arrays by ultrasonic electroplating of durable platinum black
Arcot Desai S, et al.
Frontiers in Neuroengineering, 3, 5-5 (2010)
Revealing the semiconductor-catalyst interface in buried platinum black silicon photocathodes
Aguiar JA, et al.
Journal of Material Chemistry A, 4(21), 8123-8129 (2016)
A droplet-based microfluidic electrochemical sensor using platinum-black microelectrode and its application in high sensitive glucose sensing
Gu S, et al.
Biosensors And Bioelectronics, 55, 106-112 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service