520799
Platinum black
low bulk density, ≥99.9% trace metals basis
Synonym(s):
Platinum gray, Pt, Pt Black
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥99.9% trace metals basis
form
powder
reaction suitability
core: platinum
reagent type: catalyst
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
surface area
27-36 m2/g
greener alternative category
SMILES string
[Pt]
InChI
1S/Pt
InChI key
BASFCYQUMIYNBI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.
Application
Platinum black (low bulk density) can be electroplated on implantable microelectrode arrays (MEAs) to improve the impedence of the device. It can be used as a microelectrode for the formation of a microfluidic electrochemical sensor for glucose sensing applications. It can also be used as a photocathode for the production of hydrogen.
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Buchi FN, et al. et al.
Polymer Electrolyte Fuel Cell Durability, By Felix N. Buchi, Minoru Inaba, Thomas J. Schmidt null
M. C. S. Escano, et al.,
The Journal of Physical Chemistry C, 115, 19883-19889 (2011)
Z. Liu, et al.,
ACS Applied Materials & Interfaces null
M. Ling, et al.,
International Journal of Hydrogen Energy, 36, 12833-12842 (2011)
F. Alhumaidan, et al.,
Energy and Fuels null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service