Skip to Content
Merck
All Photos(1)

Key Documents

735906

Sigma-Aldrich

Copper(I) bromide

AnhydroBeads, -10 mesh, 99.99% trace metals basis

Synonym(s):

Cuprous bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuBr
CAS Number:
Molecular Weight:
143.45
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.99% trace metals basis

form

beads

impurities

≤150.0 ppm Trace Metal Analysis

particle size

-10 mesh

mp

504 °C (lit.)

density

4.71 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Cu]Br

InChI

1S/BrH.Cu/h1H;/q;+1/p-1

InChI key

NKNDPYCGAZPOFS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cuprous bromide may be prepared by the reduction of cupric bromide or CuSO4-NaBr. In the molten state, it is grayish brown /greenish brown in color.

Application

Some reported applications of copper bromide are:
  • catalyst in cross coupling reactions.
  • co-catalyst in Sonogashira coupling.
  • Lewis acid in enantioselective addition of alkynes.
  • reducing agent, when complexed by three molecules of pyridine initiators for the controlled polymerization of styrene, methyl acrylate and methyl methacrylate.

Reductive homocoupling of α-bromo- α- chlorocarboxylates to dimethyl α, α′ dichlorosuccinate derivatives in presence of CuBr/LiOCH3 in methanol has been reported.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study on atom transfer radical polymerization initiated by p-nitrophenyl initiator and synthesis of amino end-functionalized polymers thereof
Qu, T.; Cai, W.; Fu, Z.; Shi, Y.
Macromolecular Research, 18(7), 623-629 (2010)
Shengtong Sun et al.
The journal of physical chemistry. A, 114(32), 8331-8336 (2010-08-13)
A designed ligand-accelerated Cu(I)-catalyzed cycloaddition (CuAAC) reaction was monitored for the first time by real time infrared analysis technique based on ATR-FTIR principles. Principal components analysis (PCA) and two-dimensional correlation spectroscopy (2Dcos) results showed that the consumption of alkyne and
Denissova I and Barriault L
Handbook of Reagents for Organic Synthesis null
Denissova I and Barriault L et al.
Handbook of Reagents for Organic Synthesis null
B Podobnik et al.
Bioconjugate chemistry, 26(3), 452-459 (2015-01-30)
The covalent attachment of poly(ethylene glycol) (PEG) to therapeutic proteins is a commonly used approach for extending in vivo half-lives. A potential limitation of this PEGylation strategy is the adverse effect of PEG on conjugate viscosity. Interferon-alpha (IFN) was conjugated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service