Skip to Content
Merck
All Photos(2)

Key Documents

437867

Sigma-Aldrich

Copper(II) bromide

99.999% trace metals basis

Synonym(s):

Cupric bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuBr2
CAS Number:
Molecular Weight:
223.35
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

solid

impurities

≤15.0 ppm Trace Metal Analysis

mp

498 °C (lit.)

density

4.77 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Br[Cu]Br

InChI

1S/2BrH.Cu/h2*1H;/q;;+2/p-2

InChI key

QTMDXZNDVAMKGV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Copper bromide is a monoclinic crystalline material. It is soluble in water, alcohol and acetone. It may be synthesized by neutralizing oxides, carbonates or hydroxides of copper with hydrobromic acid. Another synthesis technique is by oxidizing copper either with bromine water or bromine solutions in alcohol.

Application

Copper bromide may be used as a catalyst in organic reactions and as a brominating agent. Copper catalyzed biotinylation of acetylene terminated poly(ethylene glycol) methyl ether methylacrylate (PEGMEMA) chains has been investigated. Copper dibromide in acetonitrile acts as a catalyst in the interconversion of acetals to bis(methoxyphenyl)methyl (BMPM) ethers. It has been used as a catalyst in the intramolecular decarboxylative functionalization of α-carbonyl to yield a C(sp(3))-O bond for the synthesis of furo[3,2-c]coumarins. Poly(3,4-ethylene dioxythiophene):poly(styrenesulfonate) (PEDOT:PSS) has been reportedly doped with CuBr2 to act as hole transport layer (HTL) in polymer solar cells(PSCs). Doping increase the conductivity and thereby increasing the device power conversion efficiency of PSCs.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper(ii) bromide-catalyzed intramolecular decarboxylative functionalization to form a C(sp(3))-O bond for the synthesis of furo[3,2-c]coumarins.
Zhang WL, et al.
Organic & Biomolecular Chemistry, 13(12), 3602-3609 null
Zhiqiang Zhao et al.
ACS applied materials & interfaces, 7(3), 1439-1448 (2014-12-24)
Copper(II) bromide (CuBr2) salt has been applied to dope poly(3,4-ethylene dioxythiophene):poly(styrenesulfonate) (PEDOT:PSS) as the hole transport layer (HTL) in polymer solar cells (PSCs), improving dramatically the conductivity of PEDOT:PSS film and consequently the device power conversion efficiency (PCE). Under the
Richardson HW
Handbook of Copper Compounds and Applications null
Copper(II) bromide as an efficient catalyst for acetal to bisarylmethyl ether interconversion
Mezaache R, et al.
Tetrahedron Letters, 55(52), 7167-7171 (2014)
Gavin T Knight et al.
Journal of visualized experiments : JoVE, 92(92), e52186-e52186 (2014-11-20)
In tissue engineering, it is desirable to exhibit spatial control of tissue morphology and cell fate in culture on the micron scale. Culture substrates presenting grafted poly(ethylene glycol) (PEG) brushes can be used to achieve this task by creating microscale

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service