Skip to Content
Merck
All Photos(1)

Key Documents

724858

Sigma-Aldrich

Poly(N-isopropylacrylamide-co-methacrylic acid)

methacrylic acid 10 mol %, Mn 60,000

Synonym(s):

Poly(NIPAM-co-MAA), Polyacrylamide, functionalized polyNIPAM, functionalized polyacrylamide, polyNIPAM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11NO)m (C4H6O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

Quality Level

mol wt

Mn 60,000

composition

methacrylic acid, 10 mol %

mp

>300 °C

Mw/Mn

≤2.5

SMILES string

N(C(C)C)C(=O)C=C.OC(=O)C(=C)C

InChI

1S/C6H11NO.C4H6O2/c1-4-6(8)7-5(2)3;1-3(2)4(5)6/h4-5H,1H2,2-3H3,(H,7,8);1H2,2H3,(H,5,6)

InChI key

BGJOTKHBFYMJST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Intelligent Swelling/Collapsing copolymer that can be used as a temperature- and pH-sensitive materials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunlu Dai et al.
ACS nano, 6(4), 3327-3338 (2012-03-23)
In this study, we report a new controlled release system based on up-conversion luminescent microspheres of NaYF(4):Yb(3+)/Er(3+) coated with the smart hydrogel poly[(N-isopropylacrylamide)-co-(methacrylic acid)] (P(NIPAM-co-MAA)) (prepared using 5 mol % of MAA) shell. The hybrid microspheres show bright up-conversion fluorescence
Kai Zhang et al.
Biomacromolecules, 5(4), 1248-1255 (2004-07-13)
To elucidate the mechanism of stimuli-responsive permeability and to optimize the design, the nanostructure of polymeric composite membranes, developed in our laboratory, was characterized. The membranes were prepared to contain various amounts of stimuli-responsive nanoparticles of poly(N-isopropylacrylamide-co-methacrylic acid), with or
Kai Zhang et al.
The Journal of pharmacy and pharmacology, 56(5), 611-620 (2004-05-15)
A new glucose-responsive polymeric composite membrane that provided pulsatile insulin release was developed in our laboratory previously. To develop a clinically useful insulin delivery system, this study was designed to investigate factors influencing insulin stability during delivery by this membrane.
Pierre Simard et al.
International journal of pharmaceutics, 381(2), 86-96 (2009-05-19)
A promising avenue in cancer therapy using liposomal formulations is the combination of site-specific delivery with triggered drug release. The use of trigger mechanisms in liposomes could be relevant for drugs susceptible to lysosomal hydrolytic/enzymatic degradation. Here, we propose a
Sabrina Schmidt et al.
Langmuir : the ACS journal of surfaces and colloids, 27(16), 9801-9806 (2011-07-09)
Charged poly(N-isopropylacrylamide-co-methacrylic acid) [P(NiPAM-co-MAA)] microgels can stabilize thermo- and pH-sensitive emulsions. By placing charged units at different locations in the microgels and comparing the emulsion properties, we demonstrate that their behaviors as emulsion stabilizers are very different from molecular surfactants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service