Skip to Content
Merck
All Photos(1)

Key Documents

736236

Sigma-Aldrich

2-Cyanopropan-2-yl N-methyl-N-(pyridin-4-yl)carbamodithioate

97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3S2
CAS Number:
Molecular Weight:
251.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Assay

97% (HPLC)

form

solid

mp

97-102 °C

storage temp.

2-8°C

SMILES string

CN(C(=S)SC(C)(C)C#N)c1ccncc1

InChI

1S/C11H13N3S2/c1-11(2,8-12)16-10(15)14(3)9-4-6-13-7-5-9/h4-7H,1-3H3

InChI key

PYVIBYAVTHICFS-UHFFFAOYSA-N

Application

Switchable RAFT agent for controlled radical polymerization. The neutral form is well-suited for polymerization of vinyl esters and vinyl amides (LAMs), and the protonated form is well-suited for styrenes acrylates and methacrylates (MAMs). Chain Transfer Agent (CTA)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RAFT Agent Design and Synthesis
Keddie, D. J.; et al.
Macromolecules, 45, 5321-5342 (2012)
Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service