719978
Resomer® R 202 H, Poly(D,L-lactide)
acid terminated, Mw 10,000-18,000
Synonym(s):
Lactide polymer, PDLLA, Polylactide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
(C6H8O4)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
form
amorphous
mol wt
Mw 10,000-18,000
degradation timeframe
<6 months
viscosity
0.16-0.24 dL/g, 0.5 % (w/v) in chloroform(25 °C, Ubbelohde) (size 0c glass capillary viscometer)
transition temp
Tg 44-48 °C
storage temp.
2-8°C
SMILES string
O1C(C(=O)OC(C1=O)C)C
InChI
1S/C6H8O4/c1-3-5(7)10-4(2)6(8)9-3/h3-4H,1-2H3
InChI key
JJTUDXZGHPGLLC-UHFFFAOYSA-N
Application
Controlled release
Legal Information
Product of Evonik
RESOMER is a registered trademark of Evonik Rohm GmbH
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
nwg
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service