Skip to Content
Merck
All Photos(3)

Key Documents

739952

Sigma-Aldrich

Resomer® RG 503, Poly(D,L-lactide-co-glycolide)

lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000

Synonym(s):

PLGA, RESOMER® RG 503

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C3H4O2]x[C2H2O2]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

amorphous

feed ratio

lactide:glycolide 50:50

mol wt

Mw 24,000-38,000

degradation timeframe

<3 months

viscosity

0.32-0.44 dL/g, 0.1 % (w/v) in chloroform(25 °C, Ubbelohde) (size 0c glass capillary viscometer)

transition temp

Tg 44-48 °C

storage temp.

2-8°C

SMILES string

O2C(C(=O)OC(C2=O)C)C.O1CC(=O)OCC1=O

InChI

1S/C6H8O4.C4H4O4/c1-3-5(7)10-4(2)6(8)9-3;5-3-1-7-4(6)2-8-3/h3-4H,1-2H3;1-2H2

InChI key

LCSKNASZPVZHEG-UHFFFAOYSA-N

Application

Controlled release

Legal Information

Product of Evonik
RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaiping Wang et al.
Journal of microencapsulation, 36(4), 356-370 (2019-06-14)
Here, we have successfully synthesised and purified multifunctional PLGA-based nanoparticles by the co-encapsulation of an anticancer drug (tetrandrine) and a magnetic material (Fe3O4). The obtained Tet-Fe3O4-PLGA NPs had a uniform spherical shape with a particle size of approximately 199 nm and
William A Lackington et al.
Journal of controlled release : official journal of the Controlled Release Society, 304, 51-64 (2019-05-06)
Neurotrophic factor delivery via biodegradable nerve guidance conduits may serve as a promising treatment for the repair of large peripheral nerve defects. However, a platform for controlled delivery is required because of their short in vivo half-life and their potential
Rongcai Liang et al.
International journal of pharmaceutics, 454(1), 344-353 (2013-07-23)
Peptide or protein degradation often occurs when water flows into the dosage form. The aim of this study was to investigate the effect of water on exenatide acylation in poly(lactide-co-glycolide) (PLGA) microspheres. Exenatide-loaded PLGA microspheres were incubated at different relative
Teresa Musumeci et al.
International journal of pharmaceutics, 440(2), 135-140 (2012-10-20)
Melatonin, a neurohormone secreted by the pineal gland, is able to modulate intraocular pressure (IOP). The aim of this study was to generate nanoparticle (NPs) sustained release formulations that allow to extend the pre-corneal residence time of melatonin, thus prolonging
Igor Jeroukhimov et al.
Journal of the American College of Surgeons, 218(1), 102-107 (2013-11-12)
Chronic pain after inguinal hernia repair occurs in 16% to 62% of patients. The underlying mechanism probably involves sensory nerve damage and abnormal healing that might be influenced by the materials chosen for the procedure. We hypothesize that nonabsorbable sutures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service