Skip to Content
Merck
All Photos(1)

Key Documents

71746

Sigma-Aldrich

Sodium metasilicate pentahydrate

≥95.0% (T)

Synonym(s):

Disodium silicate pentahydrate, Sodium silicate hydrate, Uniflo 26

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SiO3 · 5H2O
CAS Number:
Molecular Weight:
212.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥95.0% (T)
form:
beads

Quality Level

Assay

≥95.0% (T)

form

beads

SMILES string

O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

InChI

1S/2Na.O3Si.5H2O/c;;1-4(2)3;;;;;/h;;;5*1H2/q2*+1;-2;;;;;

InChI key

ADPGKKZKGXANON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium metasilicate pentahydrate can be used:
  • As a silicate electrolyte in plasma electrolytic oxidation (PEO) coating.
  • For the synthesis of sintered mullite ceramics (3Al2O3·2SiO2) by the co-precipitation method.
  • As a source of silica for the synthesis of silica-zirconia composites for the chromatographic applications.
  • For the synthesis of photo-Fenton catalyst, nontronite, for the degradation of Rhodamine B.
  • As a starting material for the synthesis of zeolite NaA layers on α-Al2O3 supports.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of AZ31 Mg Alloy coated by plasma electrolytic oxidation.
Durdu S, et al.
Vacuum, 88, 130-133 (2013)
A sol-gel preparation of silica coated zirconia microspheres as chromatographic support materials.
Shalliker RA, et al.
J. Mater. Sci., 32(11), 2949-2955 (1997)
Layer development and growth history of polycrystalline zeolite A membranes synthesised from a clear solution.
Zah J, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 93(1-3), 141-150 (2006)
A novel photosensitized Fenton reaction catalyzed by sandwiched iron in synthetic nontronite.
Liu R, et al.
Royal Society of Chemistry Advances, 4(25), 12958-12963 (2014)
Mechanical, morphological and dielectric properties of sintered mullite ceramics at two different heating rates prepared from alkaline monophasic salts.
Sanad MMS, et al.
Ceramics International, 39(2), 1547-1554 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service