Skip to Content
Merck
All Photos(1)

Key Documents

208299

Sigma-Aldrich

Cadmium chloride hydrate

98%

Synonym(s):

Cadmium dichloride hydrate, Cadmium(2+) dichloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdCl2.xH2O
CAS Number:
Molecular Weight:
183.32 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor density

6.3 (vs air)

vapor pressure

10 mmHg ( 656 °C)

Assay

98%

form

solid

reaction suitability

reagent type: catalyst
core: cadmium

density

3.327 g/mL at 25 °C (lit.)

SMILES string

O.Cl[Cd]Cl

InChI

1S/Cd.2ClH.H2O/h;2*1H;1H2/q+2;;;/p-2

InChI key

OISMQLUZKQIKII-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kovo G Akpomie et al.
Scientific reports, 10(1), 17094-17094 (2020-10-15)
This work was aimed at the synthesis of a hybrid (STpe-AgNP), obtained by impregnation of silver nanoparticles (AgNP) onto Solanum tuberosum peel (STpe), for the ultrasonic assisted adsorption of bromophenol blue (BB) dye. SEM, FTIR, XRD, EDX, TGA and BET
Benedikt Wimmer et al.
Analytical and bioanalytical chemistry, 412(20), 4967-4983 (2020-06-12)
In this study, we developed and validated a CE-TOF-MS method for the quantification of glyphosate (N-(phosphonomethyl)glycine) and its major degradation product aminomethylphosphonic acid (AMPA) in different samples including beer, media from toxicological analysis with Daphnia magna, and sorption experiments. Using
Thi-Hai Anh Nguyen et al.
Waste management (New York, N.Y.), 120, 755-761 (2020-11-26)
Recycling opportunities for graphitic carbon from lithium-ion battery (LIB) anodes have been neglected owing to the relative low value of application. In this study, the potential methods for removing toxic metals (lead, barium, and cadmium) and organic compounds (2,4-dinitrotoluene [DNT]
Alan Kania et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(28), 5362-5375 (2020-06-14)
Binge-eating disorder is the most common eating disorder. Various neuropeptides play important roles in the regulation of feeding behavior, including relaxin-3 (RLN3), which stimulates food intake in rats through the activation of the relaxin-family peptide-3 receptor (RXFP3). Here we demonstrate
Po-Jung Jimmy Huang et al.
Analytical chemistry, 87(13), 6890-6895 (2015-06-11)
Mercury is a highly toxic heavy metal, and detection of Hg(2+) by biosensors has attracted extensive research interest in the past decade. In particular, a number of DNA-based sensing strategies have been developed. Well-known examples include thymine-Hg(2+) interactions and Hg(2+)-activated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service