Skip to Content
Merck
All Photos(1)

Key Documents

714984

Sigma-Aldrich

Ethyl 2,3-pentadienoate

technical grade, >85%

Synonym(s):

2,3-Pentadienoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O2
CAS Number:
Molecular Weight:
126.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

>85%

form

liquid

refractive index

n20/D 1.464

storage temp.

−20°C

SMILES string

CCOC(=O)C=C=CC

InChI

1S/C7H10O2/c1-3-5-6-7(8)9-4-2/h3,6H,4H2,1-2H3

InChI key

JGEKOPSQLCPNGA-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

124.0 °F

Flash Point(C)

51.1 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service