Skip to Content
Merck
All Photos(1)

Key Documents

494992

Sigma-Aldrich

Ethyl 2,3-butadienoate

95%

Synonym(s):

(Ethoxycarbonyl)allene, Ethyl allenecarboxylate, Ethyl butadienoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C=CHCO2C2H5
CAS Number:
Molecular Weight:
112.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

refractive index

n20/D 1.46 (lit.)

bp

80 °C/60 mmHg (lit.)

density

0.966 g/mL at 25 °C (lit.)

SMILES string

[H]C([H])=C=C([H])C(=O)OCC

InChI

1S/C6H8O2/c1-3-5-6(7)8-4-2/h5H,1,4H2,2H3

InChI key

GLSUOACRAMLJIW-UHFFFAOYSA-N

General description

Ethyl 2,3-butadienoate is an α-allenic ester. The reaction of ethyl 2,3-butadienoate with N-tosylated imines in the presence of DABCO (1,4-diazabicyclo[2.2.2]octane) or DMAP (4-dimethylaminopyridine) forms azetidine derivatives or novel dihydropyridine derivatives respectively. The performance of bifunctional N-acyl aminophosphines to catalyze the asymmetric [3+2] cycloaddition of phenylidenemalononitrile with ethyl 2,3-butadienoate has been evaluated.

Application

Ethyl 2,3-butadienoate may be used in the synthesis of dihydropyrans by reacting with acyclic enones. It may also be used to synthesize spiranic heterocycles by reacting with heterocyclic bis-arylidene ketones via phosphine-catalyzed [3+2] annulations.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Abnormal aza-Baylis-Hillman reaction of N-tosylated imines with ethyl 2,3-butadienoate and penta-3,4-dien-2-one.
Zhao GL, et al.
Organic Letters, 5(24), 4737-4739 (2003)
Asymmetric [3+2] Cycloadditions of Allenoates and Dual Activated Olefins Catalyzed by Simple Bifunctional N-Acyl Aminophosphines.
Xiao H, et al.
Angewandte Chemie (International Edition in English), 49(26), 4467-4470 (2010)
Heterocyclic Spiranes and Dispiranes via Enantioselective Phosphine-Catalyzed [3+2] Annulations.
Duvvuru D, et al.
Advanced Synthesis & Catalysis, 354(2-3), 408-414 (2012)
Development of a formal catalytic asymmetric [4+2] addition of ethyl-2,3-butadienoate with acyclic enones.
Ashtekar KD, et al.
Organic Letters, 13(21), 5732-5735 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service