Skip to Content
Merck
All Photos(4)

Documents

703508

Sigma-Aldrich

Lithium aluminum hydride solution

greener alternative

2.3 M in 2-methyltetrahydrofuran

Synonym(s):

LAH, Lithium alanate, Lithium tetrahydroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiAlH4
CAS Number:
Molecular Weight:
37.95
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: reductant

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

10 % (w/w)
2.3 M in 2-methyltetrahydrofuran

density

0.883 g/mL at 25 °C

greener alternative category

storage temp.

2-30°C

SMILES string

[Li+].[AlH4-]

InChI

1S/Al.Li.4H/q-1;+1;;;;

InChI key

OCZDCIYGECBNKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product, 2.3 M in 2-methyltetrahydrofuran aligns with Safer Solvents and Auxiliaries, Use of Renewable Feedstocks and Inherently Safer Chemistry for Accident Prevention. Click here for more information.

Application

Powerful reducing agent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

2-Methyltetrahydrofuran (2-MeTHF): A Biomass-Derived Solvent with Broad Application in Organic Chemistry
Reactant or reagent for:
  • The preparation of thermoplastic polyester polyamides from oleic acid
  • Lithium-polymer batteries
  • Hydrodefluorination of gem-difluoromethylene derivatives
  • Asymmetric aldol reactions
  • Synthesis of Li-Al-N-H composites with hydrogen absorption / desorption properties

LAH is a powerful reducing agent for many different reduction reactions such as that of ketones to alcohols

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup - Solvent

Flash Point(C)

-12 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pseudo-allosteric recognition of mandelic acid with an enantioselective coordination complex.
Jungseok Heo et al.
Angewandte Chemie (International ed. in English), 45(6), 941-944 (2005-12-31)
Sonja Stanković et al.
Organic & biomolecular chemistry, 8(19), 4266-4273 (2010-08-05)
A new synthetic protocol for the LiAlH(4)-promoted reduction of non-activated aziridines under microwave conditions was developed. Thus, ring opening of 2-(acetoxymethyl)aziridines provided the corresponding beta-amino alcohols, which were then used as eligible substrates in the synthesis of 5-methylmorpholin-2-ones via condensation
Matthias D'hooghe et al.
Organic & biomolecular chemistry, 8(3), 607-615 (2010-01-22)
trans- and cis-1-Alkyl-4-aryl-3-chloroazetidin-2-ones, prepared through cyclocondensation of chloroketene and the appropriate imines in a diastereoselective way, were transformed into the corresponding non-activated trans- and cis-2-aryl-3-(hydroxymethyl)aziridines via reductive ring contraction using LiAlH(4) in Et(2)O. Furthermore, trans-2-aryl-3-(hydroxymethyl)aziridines were transformed into 2-amino-3-arylpropan-1-ols and
Seiji Takeda et al.
Journal of biomedical nanotechnology, 9(2), 303-306 (2013-05-01)
Measurement of oxidized low-density lipoprotein (LDL) generated by oxidative stress of various kinds might be useful for evaluating the risk of cardiovascular disease. We evaluated some electrode materials to detect oxidized LDL electrochemically. Some carbon nanotube dispersions were studied as
Hideki Hashizume et al.
The Journal of antibiotics, 57(1), 52-58 (2004-03-23)
Planar structures of tripropeptins (TPPs) were elucidated by spectroscopic studies including various NMR measurements. Stereochemistry of constituent amino acids of tripropeptin C (TPPC) (3) was identified by marfey's method except hydroxyproline which was determined by studies of NMR and CD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service