Skip to Content
Merck
All Photos(2)

Documents

686034

Sigma-Aldrich

Lithium aluminum hydride

greener alternative

hydrogen-storage grade

Synonym(s):

LAH, Lithium alanate, Lithium tetrahydroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiAlH4
CAS Number:
Molecular Weight:
37.95
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

hydrogen-storage grade

Assay

>99.95%

form

pellets

reaction suitability

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

125 °C (dec.) (lit.)

greener alternative category

SMILES string

[Li].[AlH3]

InChI

1S/Al.Li.4H/q-1;+1;;;;

InChI key

OCZDCIYGECBNKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Lithium aluminum hydride (LAH) is majorly used in hydrogen storage applications. Its properties include high gravimetric and volumetric hydrogen densities. It can also be used as a reducing agent in the preparation of reduced graphene oxide (rGO).

Analysis Note

Hydrogen content, XRD plots and metal purity data are available upon request.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium aluminum hydride as reducing agent for chemically reduced graphene oxides
Ambrosi A, et al.
Chemistry of Materials, 24(12), 2292-2298 (2012)
A first-principles study of the electronic structure and stability of a lithium aluminum hydride for hydrogen storage
Song Y, et al.
The Journal of Physical Chemistry B, 110(13), 6906-6910 (2006)
Reversible hydrogen storage via titanium-catalyzed LiAlH4 and Li3AlH6
Chen J, et al.
The Journal of Physical Chemistry B, 105(45), 11214-11220 (2001)
Regeneration of lithium aluminum hydride
Graetz J, et al.
Journal of the American Chemical Society, 130(52), 17790-17794 (2008)
Pseudo-allosteric recognition of mandelic acid with an enantioselective coordination complex.
Jungseok Heo et al.
Angewandte Chemie (International ed. in English), 45(6), 941-944 (2005-12-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service