Skip to Content
Merck
All Photos(1)

Key Documents

700703

Sigma-Aldrich

N-tert-Butyl-N-(2-methyl-1-phenylpropyl)-O-(1-phenylethyl)hydroxylamine

Synonym(s):

2,2,5-Trimethyl-3-(1-phenylethoxy)-4-phenyl-3-azahexane, Alkoxyamine NMP initiator

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31NO
CAS Number:
Molecular Weight:
325.49
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

refractive index

n20/D 1.536

density

0.968 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CC(C)C(N(OC(C)c1ccccc1)C(C)(C)C)c2ccccc2

InChI

1S/C22H31NO/c1-17(2)21(20-15-11-8-12-16-20)23(22(4,5)6)24-18(3)19-13-9-7-10-14-19/h7-18,21H,1-6H3

InChI key

FPFRTDOMIFBPBZ-UHFFFAOYSA-N

General description

For a synthetic protocol using NMP initiators, contributed by Prof. Karen Wooley, please visit our technology spotlight.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benoit, D., Chaplinski, V., Braslau, R., Hawker, C.J.
Journal of the American Chemical Society, 121, 3904-3904 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service