591947
Poly(pentafluorostyrene)
Synonym(s):
2,3,4,5,6-Pentafluorostyrene homopolymer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C8H3F5)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23
Recommended Products
form
powder
transition temp
Tg 80 °C
solubility
chloroform: soluble
SMILES string
Fc1c(c(c(c(c1F)F)C=C)F)F
InChI
1S/C8H3F5/c1-2-3-4(9)6(11)8(13)7(12)5(3)10/h2H,1H2
InChI key
LVJZCPNIJXVIAT-UHFFFAOYSA-N
Application
Low refractive index polymer for cladding layer in optical waveguides.
Poly(pentafluorostyrene) (PPFS) can be used in the development of polylectrolytic membranes for fuel cell applications. It can also act as a glycosylated polymer for potential usage in biological imaging. It is a fluorinated polymer which may be utilized in microelectronics or as an antifouling and antifogging agent.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Post-modification of poly (pentafluorostyrene): a versatile ?click? method to create well-defined multifunctional graft copolymers
Ott C, et al.
Chemical Communications (Cambridge, England), 12(30), 3516-3518 (2008)
Phosphonic acid functionalized poly (pentafluorostyrene) as polyelectrolyte membrane for fuel cell application
Atanasov V, et al.
Journal of Power Sources, 343, 364-372 (2017)
Sulfonated poly (pentafluorostyrene): Synthesis \& characterization
Atanasov V, et al.
Solid State Ionics, 252, 75-83 (2013)
Uptake of Well-Defined, Highly Glycosylated, Pentafluorostyrene-Based Polymers and Nanoparticles by Human Hepatocellular Carcinoma Cells
Babiuch K, et al.
Macromolecular Bioscience, 12(9), 1190-1199 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service