Skip to Content
Merck
All Photos(1)

Key Documents

687561

Sigma-Aldrich

(Rp)-2-(tert-Butylthio)-1-(diphenylphosphino)ferrocene

98%

Synonym(s):

(1S)-1-(tert-Butylthio)-2-(diphenylphosphino)ferrocene, (R)-FeSulPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H27FePS
CAS Number:
Molecular Weight:
458.38
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

146-149 °C

functional group

phosphine
thioether

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.CC(C)(C)S[C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4

InChI

1S/C21H22PS.C5H5.Fe/c1-21(2,3)23-20-16-10-15-19(20)22(17-11-6-4-7-12-17)18-13-8-5-9-14-18;1-2-4-5-3-1;/h4-16H,1-3H3;1-5H;

InChI key

IHULVPNADBGKMY-UHFFFAOYSA-N

Application

Enantioselective allylic substitution, enantioselective 1,3-dipolar cycloaddition.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service