Skip to Content
Merck
All Photos(1)

Documents

578266

Sigma-Aldrich

1,1′-Bis(diisopropylphosphino)ferrocene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H36FeP2
CAS Number:
Molecular Weight:
418.31
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

50-52 °C (lit.)

functional group

phosphine

SMILES string

[Fe].CC(C)P([C]1[CH][CH][CH][CH]1)C(C)C.CC(C)P([C]2[CH][CH][CH][CH]2)C(C)C

InChI

1S/2C11H18P.Fe/c2*1-9(2)12(10(3)4)11-7-5-6-8-11;/h2*5-10H,1-4H3;

InChI key

LPBBEMRSRONKMN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service