670812
O-(2-Carboxyethyl)polyethylene glycol
3,000
Synonym(s):
Polyethylene glycol, Polyethylene glycol 3400 monoacid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO(CH2CH2O)nCH2CH2COOH
CAS Number:
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.22
Recommended Products
Ω-end
hydroxyl
α-end
carboxylic acid
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service