407038
Poly(ethylene glycol) bis(carboxymethyl) ether
average MN 600, cross-linking reagent amine reactive, carboxylic acid
Synonym(s):
Polyethylene glycol, Polyethylene glycol 600 diacid, Polyglycol 600 diacid
About This Item
Recommended Products
Product Name
Poly(ethylene glycol) bis(carboxymethyl) ether, average Mn 600
form
viscous liquid
mol wt
average Mn 600
reaction suitability
reagent type: cross-linking reagent
reactivity: amine reactive
density
1.191 g/mL at 25 °C
Ω-end
carboxylic acid
α-end
carboxylic acid
polymer architecture
shape: linear
functionality: homobifunctional
SMILES string
OCCO.OCC(O)=O
InChI key
SZUIEBOFAKRNDS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
572.0 °F - closed cup
Flash Point(C)
300 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service