Skip to Content
Merck
All Photos(4)

Key Documents

M4670

Sigma-Aldrich

D-Mannosamine hydrochloride

≥98% (HPLC)

Synonym(s):

2-Amino-2-deoxy-D-mannose hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Beilstein:
3914860
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

crab (shell)
shrimp shells

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white

mp

168 °C (dec.) (lit.)

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

Cl[H].N[C@@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C6H13NO5.ClH/c7-3-5(10)4(9)2(1-8)12-6(3)11;/h2-6,8-11H,1,7H2;1H/t2-,3+,4-,5-,6?;/m1./s1

InChI key

QKPLRMLTKYXDST-OHXGPSCHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Mannosamine hydrochloride can be used for the synthesis of non-natural ManNAc analogs for the expression of thiols on cell-surface sialic acids. It has been used in a study to investigate the synthesis and high-throughput screening of N-acetyl-β-hexosaminidase inhibitor libraries targeting osteoarthritis.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srinivasa-Gopalan Sampathkumar et al.
Nature protocols, 1(5), 2377-2385 (2007-04-05)
The sialic acid biosynthetic pathway in mammalian cells utilizes N-acetyl-D-mannosamine (ManNAc) as a natural metabolic precursor and has the remarkable ability to biosynthetically process non-natural ManNAc analogs. Herein, we describe a recipe-style protocol for the synthesis of the novel peracetylated
Junjie Liu et al.
The Journal of organic chemistry, 69(19), 6273-6283 (2004-09-11)
C1 Nitrogen iminocyclitols are potent inhibitors of N-acetyl-beta-hexosaminidases. Given hexosaminidases' important roles in osteoarthritis, we developed two straightforward and efficient syntheses of C1 nitrogen iminocyclitols from two readily available starting materials, D-mannosamine hydrochloride and the microbial oxidation product of fructose.
Christopher Brigham et al.
Journal of bacteriology, 191(11), 3629-3638 (2009-03-24)
We characterized the nanLET operon in Bacteroides fragilis, whose products are required for the utilization of the sialic acid N-acetyl neuraminic acid (NANA) as a carbon and energy source. The first gene of the operon is nanL, which codes for
Tal Gefen et al.
Vaccine, 28(51), 8197-8202 (2010-09-30)
Passive immunization with cross-species antibodies triggers the patient's immune response, thereby preventing repeated treatment. Mannosamine-biotin adduct (MBA) has been described as a masking agent for immunogenic reduction and here, the immunogenicity and biological activity of MBA-coated horse anti-viper venom (hsIgG)
Monica De Caroli et al.
The Plant journal : for cell and molecular biology, 65(2), 295-308 (2011-01-13)
The secretory pathway in plants involves sustained traffic to the cell wall, as matrix components, polysaccharides and proteins reach the cell wall through the endomembrane system. We studied the secretion pattern of cell-wall proteins in tobacco protoplasts and leaf epidermal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service